Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roche Diagnostics Cobas™ Liat Bordetella Control Kit

Catalog No. 21385126 Shop All Roche Diagnostics Products
Change view
Click to view available options
Quantity:
3 Sets (Positive and Negative)/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-385-126 3 Sets (Positive and Negative)/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-385-126 Supplier Roche Diagnostics Supplier No. 09857672190
Add to Cart
Add to Cart

Control kit for use with the cobas™ Liat Bordetella Assay.

  • Designed specifically for use with the cobas™ Liat Bordetella assay
  • Provides control material to support assay performance verification
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2 to 8 °C
Clia Complexity Waived
Detectable Analytes Bordatella, B. pertussis, B. parapertussis, B. holmesii
Quantity 3 Sets (Positive and Negative)/Kit
Test Time 15 min.
Dimensions (L x W x H) 6.1 x 10 x 1.4 in.
Weight (English) 0.1 lb.
Form Ready-to-Use (RTU)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.