Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roche Diagnostics Cobas™ Liat Bordetella RT-PCR Assay Kit

Catalog No. 21385125 Shop All Roche Diagnostics Products
Change view
Click to view available options
Quantity:
20 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-385-125 20 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-385-125 Supplier Roche Diagnostics Supplier No. 09857664190
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Automated in vitro nucleic acid diagnostic using real-time PCR for detection of Bordetella species in nasopharyngeal swabs.

  • Automated real-time polymerase chain reaction (PCR) test for simultaneous qualitative detection and differentiation of Bordetella pertussis (Bp), Bordetella parapertussis (Bpp), and Bordetella holmesii (Bh) nucleic acid
  • Designed for use with human nasopharyngeal swab samples from patients suspected of pertussis respiratory infection
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2 to 8 °C
Clia Complexity Waived
Detectable Analytes Bordatella, B. pertussis, B. parapertussis, B. holmesii
Sample Type Nasopharygreal Swab
Type PCR Test
Quantity 20 Tests/Kit
Sensitivity Drift B. pertussis: 99.3% PPA; B. parapertussis: 99.3% PPA; B. holmesii: 100% PPA
Test Time 15 min.
Analysis Time 15 min.
Dimensions (L x W x H) 10.7 x 6.5 x 5.7 in.
Weight (English) 2.4 lb.
Form Ready-to-Use (RTU)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.