Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roche Diagnostics Cobas™ Liat CT/NG/MG CONTROL Kit

Catalog No. 21200311 Shop All Roche Diagnostics Products
Change view
Click to view available options
Certifications/Compliance:
CE
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
21-200-311 CE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-200-311 Supplier Roche Diagnostics Supplier No. 09449639190
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cobas™ liat CT/NG/MG control kit for use with the Cobas™ liat CT/NG/MG assay or Cobas™ liat CT/NG assay.

cobas™ liat CT/NG/MG control kit for use with the cobas™ liat CT/NG/MG assay or cobas™ liat CT/NG assay.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE
Content And Storage 2°C to 8°C
Concentration Positive: 3 x 0.3 mL; Negative: 3 x 0.3 mL
Detectable Analytes Chlamydia trachomatis (CT) nucleic acids, Neisseria gonorrhoeae (NG) nucleic acids
Format 3 sets of positive and negative controls per kit
For Use With (Equipment) cobas™ liat analyzer CT/NG and CT/NG/MG assay kits
Sample Type Urine, Swab
Type IVD Control
Includes 3 sets of positive and negative controls per kit
Intended Use Control tests to be used for the cobas™ liat CT/NG and CT/NG/MG assays. The cobas™ liat CT/NG/MG test is an automated, qualitative in vitro nucleic acid diagnostic using real-time PCR for the detection of CT, NG, and MG in male urine and vaginal swabs. The cobas™ liat CT/NG test is an automated, qualitative in vitro nucleic acid diagnostic using real-time PCR for the detection of CT and NG in male urine and vaginal swabs.
Level 2
Packaging Packet
Product Name cobas™ liat CT/NG/MG control kit
Quantity 3 sets of positive and negative controls per kit
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.