Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roche Diagnostics cobas™ liat Influenza A/B + RSV Multiplex Assay

Catalog No. 23312200 Shop All Roche Diagnostics Products
Change view
Click to view available options
Quantity:
20 Tests/Cs. (Kit)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-312-200 20 Tests/Cs. (Kit)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Order eligibility is based upon executed contractual agreement.
Catalog No. 23-312-200 Supplier Roche Diagnostics Supplier No. 08160104190
 Restricted Item
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Order eligibility is based upon executed contractual agreement.

Automated multiplex real-time RT-PCR assay for the rapid in vitro qualitative detection and discrimination of Influenza A and B virus and RSV RNA in nasopharyngeal swab specimens from patients with signs and symptoms of respiratory infection.

  • No patient age restrictions for test
  • Testing accuracy with RT-PCR technology
  • Definitive results with no need for interpretation or confirmation
  • Fast, easy operation
  • Three easy steps
  • Results in 20 minutes
TRUSTED_SUSTAINABILITY

Specifications

Accuracy Influenza A: Positive Agreement 98.4%, Negative Agreement 96.5%. Influenza B: Positive Agreement 97.9%, Negative Agreement 99.4%. RSV: Positive Agreement 97.8%, Negative Agreement 98.4%
Certifications/Compliance None
Content And Storage 36°F to 46°F
Control Sets Controls purchases separately (FCAT# 23312203)
CPT Code 87631
CE Marker No
Clia Complexity Waived
Detectable Analytes Influenza A and B, Respiratory Syncytial Virus (RSV) RNA
Format Case (Kit)
Sample Type Nasopharyngeal
Sensitivity Influenza A: Positive Agreement 98.4%, Negative Agreement 96.5%. Influenza B: Positive Agreement 97.9%, Negative Agreement 99.4%. RSV: Positive Agreement 97.8%, Negative Agreement 98.4%
Type Nucleic Acid RT-PCR Influenza A, Influenza B, RSV
Packaging Type Case (kit)
Quantity 20 Tests/Cs. (Kit)
Specificity Negative Agreement: Influenza A 96.5%; Influenza B 99.4%; RSV 98.4%
Test Time 20 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.