Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roche Diagnostics cobas Liat Influenza A/B + RSV Control Kit

Catalog No. 23312203 Shop All Roche Diagnostics Products
Change view
Click to view available options
Quantity:
3 Sets of Positive and 3 Sets of Negative Controls
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-312-203 3 Sets of Positive and 3 Sets of Negative Controls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Order eligibility is based upon executed contractual agreement.
Catalog No. 23-312-203 Supplier Roche Diagnostics Supplier No. 07402686190
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Order eligibility is based upon executed contractual agreement.

For use with the cobas Liat Influenza A/B and RSV assay

  • Three sets of positive and negative controls in each kit
  • Controls used to validate each new lot of cobas Liat Influenza A/B + RSV assays
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance None
Content And Storage 36°F to 46°F
Description cobas™ Liat Flu A/B + RSV Control Kit
Detectable Analytes Liat Influenza A/B
Format Each (kit)
Includes 3 sets of positive and 3 sets of negative controls
Intended Use Used to validate each new lot of cobas™ Liat Influenza A/B + RSV assay kit
Packaging Each (kit)
Product Name cobas™ Liat Flu A/B + RSV Control Kit
Quantity 3 Sets of Positive and 3 Sets of Negative Controls
Stability 90 days
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.