Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roche Diagnostics cobas™ Liat SARS-CoV-2, Influenza A/B and RSV Control Kit

Catalog No. 21200318 Shop All Roche Diagnostics Products
Change view
Click to view available options
Quantity:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-200-318 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-200-318 Supplier Roche Diagnostics Supplier No. 09731270190
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The cobas™ liat SARS-CoV-2, Influenza A/B and RSV control kit for use with the cobas liat SARS-CoV-2, Influenza A/B and RSV assay. Control kit includes three positive and three negative controls.

The cobas™ liat SARS-CoV-2, Influenza A/B and RSV control kit for use with the cobas liat SARS-CoV-2, Influenza A/B and RSV assay. Control kit includes three positive and three negative controls.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 20 Tests/Kit; Store at 2°C to 8°C
Control Sets integrated sample processing control, positive and negative controls
CPT Code 87637
CE Marker Yes
Clia Complexity Waived
Detectable Analytes SARS-CoV-2, Influenza A, Influenza B, Rubella Virus Antibodies
Detection Method Nucleic Acid RT-PCR
Format Kit
Sample Type Nasaopharyngeal or anterior nasal
Sensitivity Influenza A, B, RSV = 100% positive agreement; SARS-CoV-2=96.7% positive agreement
Type Test
Disposable Single-use
For Use With (Application) Detection of SARS-CoV-2, Influenza A/B, and RSV
For Use With (Equipment) Used with the cobas™ liat analyzer
Product Line cobas
Quantity 1
Specificity SARS-CoV-2=96.7% negative agreement; Influenza A = 98.1% negative agreement; Influenza B = 97.7% negative agreement; RSV = 97.2% negative agreement
Test Time 20 min.
Analysis Time 20 min.
Dimensions (L x W x H) 10.6 x 6.5 x 5.4 in.
Weight (English) 2.4 lb.
Form Ready-to-Use (RTU)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.