Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roche Diagnostics cobas™ liat SARS-CoV-2, Influenza A/B and RSV PCR Assay Kit

Catalog No. 22037929 Shop All Roche Diagnostics Products
Change view
Click to view available options
Detectable Analytes:
SARS-CoV-2, Influenza A, Influenza B
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes
22-037-929 SARS-CoV-2, Influenza A, Influenza B
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-037-929 Supplier Roche Diagnostics Supplier No. 09048634190
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A rapid RT-PCR test for simultaneous qualitative detection and differentiation of SARS-CoV-2, influenza A, and influenza B nucleic acids in nasal or nasopharyngeal swabs from symptomatic patients.

  • Rapid RT-PCR test for the simultaneous qualitative detection and differentiation of SARS-CoV-2, influenza A, and influenza B nucleic acids.
  • Designed for use with nasal or nasopharyngeal swab specimens.
  • Intended for testing symptomatic patients.
  • Uses FCAT 21200318 control kit.
TRUSTED_SUSTAINABILITY

Specifications

CPT Code 87631
CE Marker CE-IVD
Clia Complexity Not Applicable
Detectable Analytes SARS-CoV-2, Influenza A, Influenza B
Format Assay Tube
Sample Type Swab
Sensitivity Covid: 94.5% PPA NPS, 96.7% PPA NS; Flu A: 100% PPA NPS, 100% PPA NS; Flu B: 100% PPA NPS; 100% PPA NS
Type Assay Kit
Disposable Single-use
For Use With (Equipment) Cobas Liat Analyzer
Quantity 20 Tests
Specificity Covid: 97.6% NPA NPS, 97.2% NPA NS; Flu A: 99.3% NPA NPS, 99.3% NPA NS; Flu B: 99.3% NPA NPS; 99.5% NPA NS
Analysis Time 20 min.
Dimensions (L x W x H) 10.7 x 6.5 x 5.5 in.
Weight (English) 2.4 lb.
Form Ready-to-Use (RTU)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.