Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roche Diagnostics cobas™ liat SARS-CoV-2 PCR Assay Kit

Catalog No. 23800534 Shop All Roche Diagnostics Products
Change view
Click to view available options
Detectable Analytes:
SARS-CoV-2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes
23-800-534 SARS-CoV-2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-800-534 Supplier Roche Diagnostics Supplier No. 09049339190
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

An automated real-time reverse transcription polymerase chain reaction (RT-PCR) test intended for the qualitative detection of severe acute respiratory syndrome coronavirus.

  • Uses FCAT 21200318 control kit.
TRUSTED_SUSTAINABILITY

Specifications

CPT Code 87635
CE Marker CE-IVD
Clia Complexity Waived
Detectable Analytes SARS-CoV-2
Format Assay Tube
Type Assay Kit
Disposable Single-use
Includes 20 Foil-Wrapped Assay Tubes, Packet Of Pipettes
PCR Method RT-PCR
Quantity 20 Tests
Shipping Condition Cold-Chain
Sterility Non-sterile
Analysis Time 20 min.
Dimensions (L x W x H) 10.7 x 6.5 x 5.5 in.
Weight (English) 2.4 lb.
Form Ready-to-Use (RTU)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.