Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roche Diagnostics cobas Liat Strep A Control Kit

Catalog No. 23312202 Shop All Roche Diagnostics Products
Change view
Click to view available options
Quantity:
3 Sets of Positive and 3 Sets of Negative Controls
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-312-202 3 Sets of Positive and 3 Sets of Negative Controls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Order eligibility is based upon executed contractual agreement.
Catalog No. 23-312-202 Supplier Roche Diagnostics Supplier No. 07402678190
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Order eligibility is based upon executed contractual agreement.

For use with the cobas Liat Strep A assay.

  • Three sets of positive and negative controls in each kit
  • Controls used to validate each new lot of cobas Liat Strep A assays
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance None
Content And Storage 36°F to 46°F
Description cobas™ Liat Strep A Control Kit
Format Each (kit)
For Use With (Equipment) cobas™ Liat Analyzer and cobas Liat Strep A Assay
Includes 3 sets of positive and 3 sets of negative controls
Intended Use Used to validate each new lot of cobas™ Liat Strep A assay kit
Packaging Each (kit)
Quantity 3 Sets of Positive and 3 Sets of Negative Controls
Stability 90 days
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.