Learn More
Description
The cobas™ PCR Media Uni Swab Sample Kit is used to collect and transport human specimens. The cobas™ PCR Media serves as a nucleic acid stabilizing transport and storage medium for human specimens. Please refer to the assay-specific Instructions for Use for claimed specimen types.
Specifications
Specifications
| Storage Requirements | 15°C to 30°C |
| For Use With (Application) | The cobas™ PCR Media Uni Swab Sample Kit is used to collect and transport human specimens. |
| Color | Clear |
| Cap Color | Yellow |
| Quantity | 100 |
| Product Type | PCR Media, Uni Swab |
| Sterility | Sterile |
| Content And Storage | 15°C to 30°C |
| Shelf Life | 24 Months |
| Certifications/Compliance | Certificate of Analysis |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.