Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roche Diagnostics Cobas™ PCR Urine Sample Kit

Catalog No. 21200308 Shop All Roche Diagnostics Products
Change view
Click to view available options
Volume (Metric):
4.3 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
21-200-308 4.3 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-200-308 Supplier Roche Diagnostics Supplier No. 05170486190
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Cobas™ PCR Urine Sample Kit is used to collect and transport urine specimens.

The cobas™ PCR Urine Sample Kit is used to collect and transport urine specimens. The cobas™ PCR Media serves as a nucleic acid stabilizing transport and storage medium for urine specimens. Refer to the assay-specific Instructions for Use for further information.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Urine Collection Kit
Closure Color Yellow
Closure Type Twist cap
Disposable Single-use
Graduated Yes
Material Plastic
Sterility Sterile
Color Clear
Dimensions (L x W x H) 13.6 x 14.7 x 8.9 in.
For Use With (Application) The cobas™ PCR Urine Sample Kit is used to collect and transport urine specimens.
For Use With (Equipment) For use with the cobas liat CT/NG and CT/NG/MG assays
Includes 100 cobase™ PCR Urine Sample Packets; each packet contains one vial of cobas™ PCR Media and one disposable transfer pipette
Latex Free Yes
No. per Pack 100
Storage Requirements 15°C to 30°C
Volume (Metric) 4.3 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.