Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roche Diagnostics cobas Liat SARS-CoV-2 Assay

Catalog No. 23312207 Shop All Roche Diagnostics Products
Change view
Click to view available options
Quantity:
20 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-312-207 20 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Order eligibility is based upon executed contractual agreement.
Catalog No. 23-312-207 Supplier Roche Diagnostics Supplier No. 09408592190
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Order eligibility is based upon executed contractual agreement.

Nucleic acid test for use on cobas Liat system is the first real-time RT-PCR test authorized for use in screening and testing asymptomatic/symptomatic individuals for SARS-CoV-2 at the point of care

  • Screen asymptomatic individuals to help prevent the silent spread of SARS-CoV-2
  • Provide answers patients need in 20 min. enabling quick actions to manage patient care
  • Offers broad strain coverage of SARS-CoV-2 variants
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EUA
Content And Storage 2°C to 8°C
Control Sets SARS-CoV-2 Control Kit used to verify each new lot of SARS-CoV-2 assay
CPT Code 87635
CE Marker Yes
Clia Complexity Waived
Detectable Analytes ORF 1a/b and N gene SARS-CoV-2
Format Assay Tube
Sample Type Nasopharyngeal swab, Mid turbinate swab, Nasal swab
Sensitivity SARS-CoV-2: Symptomatic individuals: 96.1% Positive Agreement, 96.8% Negative Agreement; Asymptomatic individuals 100% Positive Agreement, 98.9% Negative Agreement* *compared to an FDA-cleared EUA, Cobas™ SARS-CoV-2 Test on Cobas™ 6800/8800 systems
Type Assay Kit
Disposable Single-use
For Use With (Application) RT-PCR
For Use With (Equipment) Cobas™ Liat system analyzer
Material Plastic
PCR Method RT-PCR
Quantity 20 Tests/Kit
Shipping Condition Cold chain
Sterility Non-sterile
Target Organism Class Dual-target ORF 1a/b & N gene SARS-CoV-2
Test Time 20 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.