Learn More
Medchemexpress LLC HY-10493 5mg Medchemexpress, Cobicistat CAS:1004316-88-4 Purity:>98%
SDP
Catalog No. 501968236
Encompass_Preferred
Medchemexpress, HY-10493 5mg Cobicistat CAS:1004316-88-4 Cobicistat is a potent and selective inhibitor of cytochrome P450 3A (CYP3A) enzymes with IC50s of 30-285 nM. Cobicistat is a pharmacokinetic enhancer which increases the overall absorption of several HIV medications. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8236 Supplier Medchemexpress LLC Supplier No. HY104935MG
Medchemexpress, HY-10493 5mg Cobicistat CAS:1004316-88-4 Cobicistat is a potent and selective inhibitor of cytochrome P450 3A (CYP3A) enzymes with IC50s of 30-285 nM. Cobicistat is a pharmacokinetic enhancer which increases the overall absorption of several HIV medications. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.