Learn More
Medchemexpress LLC HY-10493 100mg , Cobicistat GS-9350 CAS:1004316-88-4 Purity:98%
SDP
Catalog No. 501493305
Encompass_Preferred
Medchemexpress, HY-10493 100mg Cobicistat GS-9350 CAS:1004316-88-4 Formula:C40H53N7O5S2 IC50: 30-285 nM (Cytochrome P450) Purity:98% Cobicistat is a potent, and selective inhibitor of cytochrome P450 3A (CYP3A) enzymes with IC 50 of 30-285 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3305 Supplier Medchemexpress LLC Supplier No. HY10493100MG
Medchemexpress, HY-10493 100mg Cobicistat GS-9350 CAS:1004316-88-4 Formula:C40H53N7O5S2 IC50: 30-285 nM (Cytochrome P450) Purity:98% Cobicistat is a potent, and selective inhibitor of cytochrome P450 3A (CYP3A) enzymes with IC 50 of 30-285 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.