Learn More
Apexbio Technology LLC Cobicistat (GS-9350), 10mg. Cas: 1004316-88-4 MFCD: MFCD18251449. Selective CYP3A inhibitor
SDP
Supplier:  Apexbio Technology LLC A431310
Encompass_Preferred
Cobicistat is a selective inhibitor of cytochrome P450 (CYP) 3A enzymes with IC50 values ranging from 0.03 to 0.285M [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4916