Learn More
Cayman Chemical CobImetInIb 10mg
SDP
Catalog No. 502981016
Encompass_Preferred
A selective, orally available inhibitor of MEK1 (IC50 = 4.2 nM); induces differentiation and apoptosis in cancer cells lines
Catalog No. 50-298-1016 Supplier Cayman Chemical Supplier No. 1956310 MG
A selective, orally available inhibitor of MEK1 (IC50 = 4.2 nM); induces differentiation and apoptosis in cancer cells lines