Learn More
Selleck Chemical LLC Cobimetinib
SDP
Catalog No. 5000000701
Encompass_Preferred
Cobimetinib (GDC-0973 RG7420) is a potent and highly selective MEK1 inhibitor with IC50 of 4 2 nM and showing no significant inhibition when tested against a panel of more than 100 of serine-threonine and tyrosine kinases Cobimetinib induces apoptosis Phase 3
Catalog No. 50-000-00701 Supplier Selleck Chemical LLC Supplier No. S8041100MG
Cobimetinib (GDC-0973 RG7420) is a potent and highly selective MEK1 inhibitor with IC50 of 4 2 nM and showing no significant inhibition when tested against a panel of more than 100 of serine-threonine and tyrosine kinases Cobimetinib induces apoptosis Phase 3