Learn More
Medchemexpress LLC Cobimetinib-d4 hydrochloride | 00-00-0 | MFCD22124461 | 99.3% | C21H18D4ClF3IN3O2 | 10MG
SDP
Catalog No. 5000260512
Encompass_Preferred
Cobimetinib-d4 hydrochloride is the deuterium-labeled hydrochloride salt of cobimetinib, a potent and selective MEK1 inhibitor. The labeled analog (d4) is provided for use as an analytical reference standard, internal standard in mass spectrometry, and tracer in research applications.
  • Deuterium-labeled (d4) for use as internal standard in mass spectrometry
  • Hydrochloride salt form for improved solubility
  • High purity (99.3%) suitable for analytical applications
  • Exact isotopic composition for accurate quantitation
  • Intended for research use only, not for human consumption
Catalog No. 50-002-60512 Supplier Medchemexpress LLC Supplier No. HY13064S110MG
Cobimetinib-d4 hydrochloride is the deuterium-labeled hydrochloride salt of cobimetinib, a potent and selective MEK1 inhibitor. The labeled analog (d4) is provided for use as an analytical reference standard, internal standard in mass spectrometry, and tracer in research applications.
  • Deuterium-labeled (d4) for use as internal standard in mass spectrometry
  • Hydrochloride salt form for improved solubility
  • High purity (99.3%) suitable for analytical applications
  • Exact isotopic composition for accurate quantitation
  • Intended for research use only, not for human consumption