Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cobimetinib-d4 hydrochloride | MFCD22124461 | 99.3% | 571.8 g·mol⁻¹ | C21H18D4ClF3IN3O2 | 5 MG
SDP

Catalog No. 5000262158
Encompass_Preferred

Cobimetinib-d4 hydrochloride is the deuterium-labeled hydrochloride salt of cobimetinib, supplied as a high-purity solid for research and analytical use. It is intended for use as an isotopically labeled reference standard or internal standard in assay development and quantitative analysis.

  • Deuterium-labeled hydrochloride salt for isotopic tracing and standards.
  • High reported purity: 99.3%.
  • Molecular formula C21H18D4ClF3IN3O2; molecular weight 571.8 g·mol⁻¹.
  • Supplied as a solid; follow recommended storage conditions for stability.
  • Available in small milligram quantities suitable for analytical workflows.
  • Suitable as an internal standard for LC-MS/MS and other quantitative methods.

Catalog No. 50-002-62158 Supplier Medchemexpress LLC Supplier No. HY13064S15MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cobimetinib-d4 hydrochloride is the deuterium-labeled hydrochloride salt of cobimetinib, supplied as a high-purity solid for research and analytical use. It is intended for use as an isotopically labeled reference standard or internal standard in assay development and quantitative analysis.

  • Deuterium-labeled hydrochloride salt for isotopic tracing and standards.
  • High reported purity: 99.3%.
  • Molecular formula C21H18D4ClF3IN3O2; molecular weight 571.8 g·mol⁻¹.
  • Supplied as a solid; follow recommended storage conditions for stability.
  • Available in small milligram quantities suitable for analytical workflows.
  • Suitable as an internal standard for LC-MS/MS and other quantitative methods.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.