Learn More
Medchemexpress LLC Cobimetinib racemate | 934662-91-6 | 99.4% | 531.31 g/mol | C21H21F3IN3O2 | 5 MG
SDP
Catalog No. 502081252
Encompass_Preferred
Cobimetinib racemate is the racemic analytical standard of Cobimetinib, a potent and selective MEK inhibitor used in research to probe MAPK/ERK signaling and MEK-dependent cellular responses. Supplied as a high-purity solid for analytical and laboratory applications, it is intended for use in assay development, compound characterization, and mechanistic studies.
  • Racemic analytical standard for Cobimetinib
  • Potent and selective MEK inhibitor for MAPK/ERK pathway studies
  • High purity (~99.4%) suitable for analytical applications
  • Available in small mg quantities for research use
  • Solid, white to light yellow appearance
  • Molecular weight 531.31 g/mol; formula C21H21F3IN3O2
Catalog No. 50-208-1252 Supplier Medchemexpress LLC Supplier No. HY130785MG
Cobimetinib racemate is the racemic analytical standard of Cobimetinib, a potent and selective MEK inhibitor used in research to probe MAPK/ERK signaling and MEK-dependent cellular responses. Supplied as a high-purity solid for analytical and laboratory applications, it is intended for use in assay development, compound characterization, and mechanistic studies.
  • Racemic analytical standard for Cobimetinib
  • Potent and selective MEK inhibitor for MAPK/ERK pathway studies
  • High purity (~99.4%) suitable for analytical applications
  • Available in small mg quantities for research use
  • Solid, white to light yellow appearance
  • Molecular weight 531.31 g/mol; formula C21H21F3IN3O2