Learn More
Aviva Systems Biology Corp COBLL1 Antibody - N-terminal region (ARP42423_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42423_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against COBLL1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human COBLL1. Alternate names: KIAA0977, COBLR1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58223