Learn More
Medchemexpress LLC Coblopasvir dihydrochloride | 1966138-53-3 | >=98.0% | 855.81 | C41H52Cl2N8O8 | 1 MG
SDP
Catalog No. 5000227523
Encompass_Preferred
Coblopasvir dihydrochloride is a pangenotypic non-structural protein 5A (NS5A) inhibitor research compound used to study chronic hepatitis C virus (HCV) infection. The compound is supplied as the dihydrochloride salt (CAS 1966138-53-3) with molecular weight 855.81 and formula C41H52Cl2N8O8.
  • Pangenotypic NS5A inhibitor suitable for antiviral research.
  • Intended for research on chronic hepatitis C virus infection.
  • Available in small, precise milligram quantities for laboratory studies.
  • High chemical purity for reproducible experimental results.
Catalog No. 50-002-27523 Supplier Medchemexpress LLC Supplier No. HY117411A1MG
Coblopasvir dihydrochloride is a pangenotypic non-structural protein 5A (NS5A) inhibitor research compound used to study chronic hepatitis C virus (HCV) infection. The compound is supplied as the dihydrochloride salt (CAS 1966138-53-3) with molecular weight 855.81 and formula C41H52Cl2N8O8.
  • Pangenotypic NS5A inhibitor suitable for antiviral research.
  • Intended for research on chronic hepatitis C virus infection.
  • Available in small, precise milligram quantities for laboratory studies.
  • High chemical purity for reproducible experimental results.