Learn More
Medchemexpress LLC Coblopasvir dihydrochloride | 1966138-53-3 | 98.5% | 855.81 | C41H52Cl2N8O8 | 25 MG
SDP
Catalog No. 5000227972
Encompass_Preferred
Coblopasvir dihydrochloride is a pangenotypic non-structural protein 5A (NS5A) inhibitor supplied for research use in studies of chronic hepatitis C virus infection. The compound is provided as the dihydrochloride salt and is intended for in vitro and preclinical research applications.
  • Pangenotypic NS5A inhibitor for HCV research.
  • Supplied as the dihydrochloride salt for improved stability and handling.
  • High purity (≈98.5%) suitable for analytical and biological studies.
  • Available in multiple pack sizes, including 25 mg and solution formats.
  • Delivered with batch-specific certificate of analysis.
Catalog No. 50-002-27972 Supplier Medchemexpress LLC Supplier No. HY117411A25MG
Coblopasvir dihydrochloride is a pangenotypic non-structural protein 5A (NS5A) inhibitor supplied for research use in studies of chronic hepatitis C virus infection. The compound is provided as the dihydrochloride salt and is intended for in vitro and preclinical research applications.
  • Pangenotypic NS5A inhibitor for HCV research.
  • Supplied as the dihydrochloride salt for improved stability and handling.
  • High purity (≈98.5%) suitable for analytical and biological studies.
  • Available in multiple pack sizes, including 25 mg and solution formats.
  • Delivered with batch-specific certificate of analysis.