Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Coblopasvir dihydrochloride | 1966138-53-3 | 99.2% | 855.81 | C41H52Cl2N8O8 | 1 ML
SDP

Catalog No. 5000226784
Encompass_Preferred

Coblopasvir dihydrochloride is the dihydrochloride salt of coblopasvir, a pan-genotypic inhibitor of hepatitis C virus nonstructural protein 5A (NS5A). It is supplied for in vitro research use as a 10 mM solution in DMSO (1 mL) or as solid plus solvent, with a reported purity of about 99.2% and a molecular weight of 855.81 Da.

  • Pangenotypic NS5A inhibitor for HCV research
  • Supplied as 10 mM in DMSO, 1 mL
  • White to off-white solid, dihydrochloride salt
  • Purity 99.19% (as stated by manufacturer)
  • Recommended storage: solid 4°C; in solvent -80°C (6 months) or -20°C (1 month)

Catalog No. 50-002-26784 Supplier Medchemexpress LLC Supplier No. HY117411A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Coblopasvir dihydrochloride is the dihydrochloride salt of coblopasvir, a pan-genotypic inhibitor of hepatitis C virus nonstructural protein 5A (NS5A). It is supplied for in vitro research use as a 10 mM solution in DMSO (1 mL) or as solid plus solvent, with a reported purity of about 99.2% and a molecular weight of 855.81 Da.

  • Pangenotypic NS5A inhibitor for HCV research
  • Supplied as 10 mM in DMSO, 1 mL
  • White to off-white solid, dihydrochloride salt
  • Purity 99.19% (as stated by manufacturer)
  • Recommended storage: solid 4°C; in solvent -80°C (6 months) or -20°C (1 month)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.