Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Coblopasvir dihydrochloride | 1966138-53-3 | 99.2% | 855.81 | C41H52Cl2N8O8 | 50 MG
SDP

Catalog No. 5000228317
Encompass_Preferred

Coblopasvir dihydrochloride is a pangenotypic non-structural protein 5A (NS5A) inhibitor provided for research use in studies of chronic hepatitis C virus (HCV) infection. The dihydrochloride salt is supplied with high purity and is available as a solid and as ready-to-use DMSO solutions for in vitro and preclinical research.

  • High purity (99.2%).
  • Molecular weight 855.81 g/mol.
  • Empirical formula C41H52Cl2N8O8.
  • Available in solid (mg quantities) and 10 mM DMSO solution formats.
  • CAS number 1966138-53-3 for unambiguous identification.
  • Store sealed, away from moisture and light; in solvent: -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-28317 Supplier Medchemexpress LLC Supplier No. HY117411A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Coblopasvir dihydrochloride is a pangenotypic non-structural protein 5A (NS5A) inhibitor provided for research use in studies of chronic hepatitis C virus (HCV) infection. The dihydrochloride salt is supplied with high purity and is available as a solid and as ready-to-use DMSO solutions for in vitro and preclinical research.

  • High purity (99.2%).
  • Molecular weight 855.81 g/mol.
  • Empirical formula C41H52Cl2N8O8.
  • Available in solid (mg quantities) and 10 mM DMSO solution formats.
  • CAS number 1966138-53-3 for unambiguous identification.
  • Store sealed, away from moisture and light; in solvent: -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.