Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Coblopasvir dihydrochloride | 1966138-53-3 | 99.2% | 855.81 g/mol | C41H52Cl2N8O8 | 100 MG
SDP

Catalog No. 5000226725
Encompass_Preferred

Coblopasvir dihydrochloride is a pangenotypic non-structural protein 5A (NS5A) inhibitor supplied as the dihydrochloride salt for laboratory research on chronic hepatitis C virus (HCV) infection.

  • Pangenotypic NS5A inhibitor for HCV research.
  • Solid, white to off-white appearance.
  • Purity 99.19%.
  • Molecular weight 855.81 g/mol; formula C41H52Cl2N8O8.
  • Available in multiple sizes including 100 MG; store sealed, away from moisture and light.

Catalog No. 50-002-26725 Supplier Medchemexpress LLC Supplier No. HY117411A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Coblopasvir dihydrochloride is a pangenotypic non-structural protein 5A (NS5A) inhibitor supplied as the dihydrochloride salt for laboratory research on chronic hepatitis C virus (HCV) infection.

  • Pangenotypic NS5A inhibitor for HCV research.
  • Solid, white to off-white appearance.
  • Purity 99.19%.
  • Molecular weight 855.81 g/mol; formula C41H52Cl2N8O8.
  • Available in multiple sizes including 100 MG; store sealed, away from moisture and light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.