Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Coblopasvir dihydrochloride | 1966138-53-3 | 99.2% | 855.81 g·mol⁻¹ | C41H52Cl2N8O8 | 5 MG
SDP

Catalog No. 5000227650
Encompass_Preferred

Coblopasvir dihydrochloride is a pangenotypic non-structural protein 5A (NS5A) inhibitor supplied as a solid research compound for preclinical studies of hepatitis C virus (HCV) replication and NS5A function. It is provided in a small research quantity with high reported purity and recommended storage conditions to maintain stability.

  • Pangenotypic NS5A inhibitor for HCV research.
  • Supplied as a solid research reagent for biochemical and virology studies.
  • High reported purity (99.19%) to support experimental consistency.
  • Chemical formula C41H52Cl2N8O8; molecular weight 855.81 g·mol⁻¹.
  • Recommended storage: 4°C sealed; in solution: -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-27650 Supplier Medchemexpress LLC Supplier No. HY117411A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Coblopasvir dihydrochloride is a pangenotypic non-structural protein 5A (NS5A) inhibitor supplied as a solid research compound for preclinical studies of hepatitis C virus (HCV) replication and NS5A function. It is provided in a small research quantity with high reported purity and recommended storage conditions to maintain stability.

  • Pangenotypic NS5A inhibitor for HCV research.
  • Supplied as a solid research reagent for biochemical and virology studies.
  • High reported purity (99.19%) to support experimental consistency.
  • Chemical formula C41H52Cl2N8O8; molecular weight 855.81 g·mol⁻¹.
  • Recommended storage: 4°C sealed; in solution: -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.