Learn More
Medchemexpress LLC Coblopasvir dihydrochloride | 1966138-53-3 | MFCD00007349 | 99.2% | 855.81 | C41H52Cl2N8O8 | 10 MG
SDP
Catalog No. 5000226264
Encompass_Preferred
Coblopasvir dihydrochloride is a research-grade pangenotypic inhibitor of hepatitis C virus non-structural protein 5A (NS5A), supplied as the dihydrochloride salt for laboratory studies on chronic HCV infection.
  • High purity suitable for research use.
  • Targets NS5A for antiviral mechanism studies.
  • Provided as the dihydrochloride salt to improve handling and stability.
  • Available in small, milligram-scale quantities for preclinical and in vitro experiments.
  • Molecular formula and weight documented for experimental planning.
Catalog No. 50-002-26264 Supplier Medchemexpress LLC Supplier No. HY117411A10MG
Coblopasvir dihydrochloride is a research-grade pangenotypic inhibitor of hepatitis C virus non-structural protein 5A (NS5A), supplied as the dihydrochloride salt for laboratory studies on chronic HCV infection.
  • High purity suitable for research use.
  • Targets NS5A for antiviral mechanism studies.
  • Provided as the dihydrochloride salt to improve handling and stability.
  • Available in small, milligram-scale quantities for preclinical and in vitro experiments.
  • Molecular formula and weight documented for experimental planning.