Learn More
Medchemexpress LLC Cobomarsen | 1848257-52-2 | C148H177N52O77P13S13 | 10MG
SDP
Catalog No. 5000261517
Encompass_Preferred
Cobomarsen is a locked nucleic acid (LNA)-based antisense oligonucleotide that selectively inhibits microRNA-155 (miR-155). Supplied as a solid (white to off-white) research reagent, it is used in preclinical studies to modulate miR-155-driven signaling and investigate oncology models by affecting pathways such as JAK/STAT, MAPK/ERK, and PI3K/AKT. The sodium salt form offers improved stability; the compound is highly water soluble and requires sealed, moisture-free storage.
  • Selective inhibition of miR-155 and related survival pathways
  • Suitable for preclinical oncology and lymphoma research
  • High aqueous solubility; may require ultrasonication
  • Improved stability as the sodium salt compared with the free acid
  • Store sealed and protected from moisture; long-term storage in solution at -80 °C
Catalog No. 50-002-61517 Supplier Medchemexpress LLC Supplier No. HY13260110MG
Cobomarsen is a locked nucleic acid (LNA)-based antisense oligonucleotide that selectively inhibits microRNA-155 (miR-155). Supplied as a solid (white to off-white) research reagent, it is used in preclinical studies to modulate miR-155-driven signaling and investigate oncology models by affecting pathways such as JAK/STAT, MAPK/ERK, and PI3K/AKT. The sodium salt form offers improved stability; the compound is highly water soluble and requires sealed, moisture-free storage.
  • Selective inhibition of miR-155 and related survival pathways
  • Suitable for preclinical oncology and lymphoma research
  • High aqueous solubility; may require ultrasonication
  • Improved stability as the sodium salt compared with the free acid
  • Store sealed and protected from moisture; long-term storage in solution at -80 °C