Learn More
Medchemexpress LLC Cobomarsen (MRG-106) | 1848257-52-2 | 4736.00 g/mol | C148H177N52O77P13S13 | 5 MG
SDP
Catalog No. 5000262307
Encompass_Preferred
Cobomarsen (MRG-106) is a locked nucleic acid-modified oligonucleotide inhibitor of microRNA-155 (miR-155) used in preclinical research to reduce cellular proliferation and survival by modulating multiple signaling pathways. The material is supplied as a solid research-grade compound and should be handled per the manufacturer's storage and handling guidance.
  • Inhibits miR-155 to modulate JAK/STAT, MAPK/ERK, and PI3K/AKT signaling.
  • Designed as an LNA/modified oligonucleotide for increased affinity and stability.
  • Provided as a white to off-white solid suitable for laboratory use.
  • CAS 1848257-52-2; molecular weight approximately 4736.00 g/mol.
  • Intended for preclinical and mechanistic studies in hematologic and lymphoid disease models.
Catalog No. 50-002-62307 Supplier Medchemexpress LLC Supplier No. HY1326015MG
Cobomarsen (MRG-106) is a locked nucleic acid-modified oligonucleotide inhibitor of microRNA-155 (miR-155) used in preclinical research to reduce cellular proliferation and survival by modulating multiple signaling pathways. The material is supplied as a solid research-grade compound and should be handled per the manufacturer's storage and handling guidance.
  • Inhibits miR-155 to modulate JAK/STAT, MAPK/ERK, and PI3K/AKT signaling.
  • Designed as an LNA/modified oligonucleotide for increased affinity and stability.
  • Provided as a white to off-white solid suitable for laboratory use.
  • CAS 1848257-52-2; molecular weight approximately 4736.00 g/mol.
  • Intended for preclinical and mechanistic studies in hematologic and lymphoid disease models.