Learn More
MSC Cobra Systems Vinyl Tape, 2 in, 150 ft, 1 Roll
Catalog No. 5000795521
Encompass
The VnM Premium Vinyl Tape is ideal for creating durable labels and signs for diverse indoor and outdoor environments, including harsh marine conditions. Its robust construction and strong adhesive ensure signs remain securely in place for years, enhancing workplace safety and productivity.
  • Offers 5-7 year outdoor life
  • Strong adhesive backing
  • Weather proven
  • Easy-to-use application
  • Features 150 ft roll length
  • Compatible with VNM4PRO printers
Catalog No. 50-007-95521 Supplier MSC Supplier No. 41603093
The VnM Premium Vinyl Tape is ideal for creating durable labels and signs for diverse indoor and outdoor environments, including harsh marine conditions. Its robust construction and strong adhesive ensure signs remain securely in place for years, enhancing workplace safety and productivity.
  • Offers 5-7 year outdoor life
  • Strong adhesive backing
  • Weather proven
  • Easy-to-use application
  • Features 150 ft roll length
  • Compatible with VNM4PRO printers