Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobra Toxin, MP Biomedicals™

Catalog No. MP219494404 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
4 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219494404 4 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219494404 Supplier MP Biomedicals, Inc Supplier No. 0219494404
Add to Cart
Add to Cart

Cobra Toxin

  • From Naja n. kaouthia
  • Purified
  • Neurotoxin.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12584-83-7
Molecular Formula C277H443N97O98S8
Molecular Weight (g/mol) 6956.65
InChI Key BVGLZNQZEYAYBJ-QWZQWHGGSA-N

Specifications

Quantity 4 mg
UN Number UN3462
Physical Form Solid
RTECSNumber GG4220000
storageNote1 Store at 2-8C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.