Learn More
Sigma Aldrich Fine Chemicals Biosciences Cocaethylene-D8 solution 1
Catalog No. 502897660
Encompass
An analog of benzoylecgonine cocaethylene is a metabolite of cocaine formed in the presence of ethanol. This stable-labeled internal standard is suitable for quantitation of cocaethylene levels in urine blood and saliva by LC/MS or GC/MS for urine drug testing clinical toxicology forensic analysis or isotope dilution methods.
Catalog No. 50-289-7660 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C0241ML
An analog of benzoylecgonine cocaethylene is a metabolite of cocaine formed in the presence of ethanol. This stable-labeled internal standard is suitable for quantitation of cocaethylene levels in urine blood and saliva by LC/MS or GC/MS for urine drug testing clinical toxicology forensic analysis or isotope dilution methods.