Learn More
Medchemexpress LLC Cochlin/COCH Human 100ug
SDP
Catalog No. 5000178493
Encompass_Preferred
Cochlin/COCH protein is involved in regulating cell shape and motility in the trabecular meshwork It exists as a monomer and may form homodimers Additionally Cochlin interacts with type II collagen SLC44A2 and ANXA2 Cochlin/COCH Protein Human (HEK293 His) is the recombinant human-derived Cochlin/COCH protein expressed by HEK293 with N-His labeled tag
Catalog No. 50-001-78493 Supplier Medchemexpress LLC Supplier No. HYP74235100UG
Cochlin/COCH protein is involved in regulating cell shape and motility in the trabecular meshwork It exists as a monomer and may form homodimers Additionally Cochlin interacts with type II collagen SLC44A2 and ANXA2 Cochlin/COCH Protein Human (HEK293 His) is the recombinant human-derived Cochlin/COCH protein expressed by HEK293 with N-His labeled tag