Learn More
Cayman Chemical CochlIoquInon A 2.5mg
SDP
Catalog No. 502981887
Encompass_Preferred
An inhibitor of DGK (Ki = 3.1 µM) and DGAT (IC50 = 5.6 µM); reduces the concentration of phosphatidic acid in T cell lymphoma (IC50 = 3 µM); binds to human CCR5 chemokine receptors with an IC50 value of 11 µM
Catalog No. 50-298-1887 Supplier Cayman Chemical Supplier No. 194402.5 MG
An inhibitor of DGK (Ki = 3.1 µM) and DGAT (IC50 = 5.6 µM); reduces the concentration of phosphatidic acid in T cell lymphoma (IC50 = 3 µM); binds to human CCR5 chemokine receptors with an IC50 value of 11 µM