Learn More
Cayman Chemical Cochlioquinone B, Purity: ≥98%, CAS: 32450-26-3, 2.5 mg
SDP
Catalog No. 502935308
Encompass_Preferred
A fungal metabolite; inhibits root growth of finger millet (E. coracana) and rice plants (O. sativa) by 59.9 and 51.7%, respectively, at 100 ppm; inhibits NADH oxidase and NADH-2,3-dimethoxy-5-pentyl-1,4-benzoquinone reductase from bovine heart mitochondria
Catalog No. 50-293-5308 Supplier Cayman Chemical Supplier No. 251212.5 MG
A fungal metabolite; inhibits root growth of finger millet (E. coracana) and rice plants (O. sativa) by 59.9 and 51.7%, respectively, at 100 ppm; inhibits NADH oxidase and NADH-2,3-dimethoxy-5-pentyl-1,4-benzoquinone reductase from bovine heart mitochondria