Learn More
AURORA BIOLABS LLC Cockroach german (bla g 2)
Catalog No. NC4007703
Encompass_Preferred
This recombinant protein contains full length of Bla g 2, a major allergen of German Cockroach, with a MW of 37.5 kDa. It is purified from HEK293 cells with >95% purity by SDS PAGE.
Catalog No. NC4007703 Supplier AURORA BIOLABS LLC Supplier No. ABRA0064M100
This recombinant protein contains full length of Bla g 2, a major allergen of German Cockroach, with a MW of 37.5 kDa. It is purified from HEK293 cells with >95% purity by SDS PAGE.