Learn More
Medchemexpress LLC (+)-Coclaurine hydrochloride | 19894-19-0 | 99.1% | 321.80 | 1 MG
SDP
Catalog No. 5000365316
Encompass_Preferred
(+)-Coclaurine hydrochloride is a benzyltetrahydroisoquinoline alkaloid isolated from various plant sources. It exhibits anti-aging activity. In in-vivo studies, this compound has been shown to influence levels of 3,4-dihydroxyphenylacetic acid and homovanillic acid in the mouse striatum, and acts as a blocker of postsynaptic dopamine receptors. It has a molecular weight of 321.80 and an appearance of a white to off-white solid.
  • Isolated from various plant sources
  • Exhibits anti-aging activity
  • Influences levels of 3,4-dihydroxyphenylacetic acid and homovanillic acid in the mouse striatum
  • Blocks postsynaptic dopamine receptors
  • White to off-white solid appearance
  • Soluble in DMSO and H2O
Catalog No. 50-003-65316 Supplier Medchemexpress LLC Supplier No. 1MGHYN2550A
(+)-Coclaurine hydrochloride is a benzyltetrahydroisoquinoline alkaloid isolated from various plant sources. It exhibits anti-aging activity. In in-vivo studies, this compound has been shown to influence levels of 3,4-dihydroxyphenylacetic acid and homovanillic acid in the mouse striatum, and acts as a blocker of postsynaptic dopamine receptors. It has a molecular weight of 321.80 and an appearance of a white to off-white solid.
  • Isolated from various plant sources
  • Exhibits anti-aging activity
  • Influences levels of 3,4-dihydroxyphenylacetic acid and homovanillic acid in the mouse striatum
  • Blocks postsynaptic dopamine receptors
  • White to off-white solid appearance
  • Soluble in DMSO and H2O