Learn More
TARGETMOL CHEMICALS INC Coclaurine hydrochloride 1MG
SDP
Catalog No. 502880224
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. (-)-Coclaurine hydrochloride is an antagonist of nAChRs isolated from Sarcopetalum harveyanum. Purity 97.75%
Catalog No. 50-288-0224 Supplier TARGETMOL CHEMICALS INC Supplier No. TN1519L1MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. (-)-Coclaurine hydrochloride is an antagonist of nAChRs isolated from Sarcopetalum harveyanum. Purity 97.75%