Learn More
Medchemexpress LLC Cocoa butter | 8002-31-1 | 100 G
SDP
Catalog No. 5000415952
Encompass_Preferred
Cocoa butter can be used as an excipient, such as lubricants and suppository bases. Pharmaceutical excipients, or pharmaceutical auxiliaries, are chemical substances used in the pharmaceutical process other than pharmaceutical ingredients. They are generally inactive ingredients in pharmaceutical preparations, which can improve the stability, solubility, and processability of pharmaceutical preparations. Pharmaceutical excipients also affect the absorption, distribution, metabolism, and elimination (ADME) processes of co-administered drugs.
  • Used as excipient (lubricants, suppository bases)
  • Improves stability, solubility and processability of pharmaceutical preparations
  • Affects ADME processes of co-administered drugs
Catalog No. 50-004-15952 Supplier Medchemexpress LLC Supplier No. HYN11684100G
Cocoa butter can be used as an excipient, such as lubricants and suppository bases. Pharmaceutical excipients, or pharmaceutical auxiliaries, are chemical substances used in the pharmaceutical process other than pharmaceutical ingredients. They are generally inactive ingredients in pharmaceutical preparations, which can improve the stability, solubility, and processability of pharmaceutical preparations. Pharmaceutical excipients also affect the absorption, distribution, metabolism, and elimination (ADME) processes of co-administered drugs.
  • Used as excipient (lubricants, suppository bases)
  • Improves stability, solubility and processability of pharmaceutical preparations
  • Affects ADME processes of co-administered drugs