Learn More
Medchemexpress LLC Cocoa butter | 8002-31-1 | 50 G
SDP
Catalog No. 5000414619
Encompass_Preferred
Cocoa butter can be used as an excipient, such as lubricants and suppository bases. Pharmaceutical excipients, or pharmaceutical auxiliaries, are chemical substances used in the pharmaceutical process other than pharmaceutical ingredients. They generally refer to inactive ingredients in pharmaceutical preparations that can improve the stability, solubility, and processability of pharmaceutical preparations. Pharmaceutical excipients also affect the absorption, distribution, metabolism, and elimination (ADME) processes of co-administered drugs.
  • Oil appearance
  • White to off-white color
Catalog No. 50-004-14619 Supplier Medchemexpress LLC Supplier No. HYN1168450G
Add to Cart
Add to Cart
Cocoa butter can be used as an excipient, such as lubricants and suppository bases. Pharmaceutical excipients, or pharmaceutical auxiliaries, are chemical substances used in the pharmaceutical process other than pharmaceutical ingredients. They generally refer to inactive ingredients in pharmaceutical preparations that can improve the stability, solubility, and processability of pharmaceutical preparations. Pharmaceutical excipients also affect the absorption, distribution, metabolism, and elimination (ADME) processes of co-administered drugs.
  • Oil appearance
  • White to off-white color