Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cocoa Butter, MP Biomedicals™

Catalog No. MP290541790 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP290541790 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP290541790 Supplier MP Biomedicals, Inc Supplier No. 0290541790
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cocoa Butter, Waxy Solid

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8002-31-1
Synonym Cacao Oil, Cocao bean oil, Cocoa absolute, Cocoa bean extract

Specifications

Melting Point Slip Melting Point: 31.7°C; Clear Melting Point: 32.1°C (literature)
Refractive Index 1.4578 (40°C)(Lit.)
Quantity 500 g
Physical Form Waxy Solid
storageNote1 Store at Room Temperature (15-30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.