Learn More
Skc West Inc Anasorb CSC, Coconut Charcoal Sorbent Tube, 50/pack
SDP
Catalog No. NC1641100
Encompass
This sorbent has a high surface area for sampling a broad range of primarily non-polar compounds. Only ships to Alaska, Arizona, California, Colorado, Hawaii, Idaho, Nevada, Oregon, Utah, Washington. Wyoming
Catalog No. NC1641100 Supplier Skc West Inc Supplier No. 22601
This sorbent has a high surface area for sampling a broad range of primarily non-polar compounds. Only ships to Alaska, Arizona, California, Colorado, Hawaii, Idaho, Nevada, Oregon, Utah, Washington. Wyoming