Learn More
SKC Inc Sorbent Tube, Anasorb CSC, Coconut Charcoal, 6 x 70-mm size, 2 sections, 50/100 mg sorbent, 20/40 mesh, with GS ends and WWW separators, fits Type A tube cover, pk/50
SDP
Catalog No. NC9362930
Encompass
SKC offers a complete selection of sorbent tubes designed specifically for solvent extraction specified in many OSHA, NIOSH, ASTM, EPA, and non-agency methods. Quality SKC Sorbent Tubes feature consistent sorbent mesh size, validation, and over 45 years of proven performance. DOES NOT ship to Alaska, Arizona, California, Colorado, Hawaii, Idaho, Nevada, Oregon, Utah, Washington. Wyoming”
Catalog No. NC9362930 Supplier SKC Inc Supplier No. 22601
SKC offers a complete selection of sorbent tubes designed specifically for solvent extraction specified in many OSHA, NIOSH, ASTM, EPA, and non-agency methods. Quality SKC Sorbent Tubes feature consistent sorbent mesh size, validation, and over 45 years of proven performance. DOES NOT ship to Alaska, Arizona, California, Colorado, Hawaii, Idaho, Nevada, Oregon, Utah, Washington. Wyoming”