Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coconut Oil, Hydrogenated, MP Biomedicals™

Catalog No. MP290140425 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 lb.
25 lb.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP290140425 25 lb.
MP290140405 5 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP290140425 Supplier MP Biomedicals, Inc Supplier No. 0290140425
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Coconut Oil, Hydrogenated, White Semi-solid

  • Coconut oil is a common ingredient of animal diet.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Synonym Coconut fat, Coconut oil from Cocos nucifera, Copra oil

Specifications

Melting Point 77.0°F (literature)
Color White
Quantity 25 lb.
Physical Form Semi-solid
storageNote1 Store at Room Temperature (15-30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.