Learn More
Electron Microscopy Sciences Cocoon Sample Box BE3D
SDP
Catalog No. 5031082
Encompass_Preferred
The Revolutionary Way for Mailing and Storing Samples Boxes - Made from polypropylene, envelope shape, available with or without an inner frame. These boxes are the safest way for mailing or storing fragile samples. They are impact and shock resistant With a living-hinge, opaque-clear, reusable and recyclable. The boxes are available in many sizes and styles to fit your needs. Closing Clips and Security Seals are also available as an option. With 6 round cavities 1.29x0.62" for storing vials DDB30, OD: 5.31x3.89x0.70", ID: 4.33x2
Catalog No. 50-310-82 Supplier Electron Microscopy Sciences Supplier No. 643003D
The Revolutionary Way for Mailing and Storing Samples Boxes - Made from polypropylene, envelope shape, available with or without an inner frame. These boxes are the safest way for mailing or storing fragile samples. They are impact and shock resistant With a living-hinge, opaque-clear, reusable and recyclable. The boxes are available in many sizes and styles to fit your needs. Closing Clips and Security Seals are also available as an option. With 6 round cavities 1.29x0.62" for storing vials DDB30, OD: 5.31x3.89x0.70", ID: 4.33x2