Learn More
SPEX CERTIPREP LLC 3P COD CRM, 500 mL
Catalog No. NC0933453
Encompass_Preferred
(NSI by Zeptometrix) COD CRM, 500 mL, 10000 mg/L in Water.
Catalog No. NC0933453 Supplier SPEX CERTIPREP LLC 3P Supplier No. IS001H500
(NSI by Zeptometrix) COD CRM, 500 mL, 10000 mg/L in Water.