Learn More
Hach Company COD2 Digestion Vials, HR, Mercury-free, pk/150
Supplier:  Hach Company 2565115
Encompass
For high range, mercury-free Chemical Oxygen Demand (COD) determinations. COD2 Digestion Vials, Mercury-Free Formulation. Range: 20-1500 mg/L COD. Pack of 150 vials
Catalog No. 50-199-6240