Learn More
Hach Company COD2 Digestion Vials, UHR, Mercury-free, pk/25
Supplier:  Hach Company 2834325
Encompass
For Ultra-High range, mercury-free chemical oxygen demand determinations. COD2 Digestion Vials, Mercury-Free Formulation. Method: Reactor Digestion. Range: 200-15,000 mg/L COD. Pack of 25 vials
Catalog No. 50-199-6744