Learn More
Hach Company COD Digestion Vials, High Range, pk/150
Supplier:  Hach Company 2125915
Encompass
For determination of high range Chemical Oxygen Demand (COD) by the Reactor Digestion method. US EPA approved for wastewater analysis using Hach Method 8000. Range: 20 -1500 mg/L COD. Pack of 150 vials.
Catalog No. 50-199-5461